Assay Details
| Gene Symbol: LOC114116722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973912.1 | XP_027829713.1 | 3 - 4 | 90 | 107 | |
| Gene Symbol: LOC101122608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973511.1 | XP_027829312.1 | 8 - 9 | 636 | 107 | |
| GenBank mRNA | FJ958364.1 | - | 8 - 9 | 435 | 107 | |
More Information
Set Membership: |
|

LOC114116722
Genomic Map