Assay Details
Gene Symbol: LOC114116722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973912.1 | XP_027829713.1 | 4 - 5 | 187 | 184 |
Gene Symbol: LOC101122608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973511.1 | XP_027829312.1 | 9 - 10 | 733 | 184 | |
GenBank mRNA | FJ958364.1 | - | 9 - 10 | 532 | 184 |

More Information
Set Membership: |
![]() ![]() |
