Assay Details
Gene Symbol: LOC101115787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968374.1 | XP_027824175.1 | 17 - 18 | 1692 | 105 |
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968627.1 | XP_027824428.1 | 7 - 8 | 1574 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
