Assay Details
Gene Symbol: RALYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184188.3 | XP_012039578.2 | 6 - 7 | 699 | 71 | |
XM_027973257.1 | XP_027829058.1 | 7 - 8 | 1745 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
