Assay Details
Gene Symbol: HOXC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094079.2 | XP_014949565.2 | 1 - 2 | 604 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
