Assay Details
Gene Symbol: LOC101115853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962283.1 | XP_027818084.1 | 5 - 6 | 758 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
