Assay Details
| Gene Symbol: ADA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006971.4 | XP_004007020.4 | 4 - 5 | 846 | 68 | |
| XM_012175611.3 | XP_012031001.3 | 4 - 5 | 846 | 68 | ||
| XM_027968053.1 | XP_027823854.1 | 4 - 5 | 827 | 68 | ||
More Information
Set Membership: |
|

ADA2
Genomic Map