Assay Details
Gene Symbol: NDUFB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008468.4 | XP_004008517.2 | 2 - 3 | 361 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
