Assay Details
| Gene Symbol: PRDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011238.4 | XP_004011287.2 | 4 - 5 | 953 | 68 | |
| XM_012182705.3 | XP_012038095.2 | 4 - 5 | 1029 | 68 | ||
| XM_027972405.1 | XP_027828206.1 | 4 - 5 | 953 | 68 | ||
More Information
Set Membership: |
|

PRDM1
Genomic Map