Assay Details
| Gene Symbol: EIF2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097694.2 | XP_011953084.1 | 6 - 7 | 672 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF2B1| Gene Symbol: EIF2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097694.2 | XP_011953084.1 | 6 - 7 | 672 | 68 | |
Set Membership: |
|