Assay Details
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959652.1 | XP_027815453.1 | 6 - 7 | 492 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
