Assay Details
Gene Symbol: LOC101115931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961952.1 | XP_027817753.1 | 13 - 14 | 4365 | 89 |
Gene Symbol: ABCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962098.1 | XP_027817899.1 | 16 - 17 | 3293 | 89 | |
XM_027962099.1 | XP_027817900.1 | 17 - 18 | 2777 | 89 | ||
XM_027962100.1 | XP_027817901.1 | 16 - 17 | 3289 | 89 | ||
XM_027962101.1 | XP_027817902.1 | 16 - 17 | 3262 | 89 | ||
XM_027962102.1 | XP_027817903.1 | 16 - 17 | 3291 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
