Assay Details
| Gene Symbol: MRPL55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008395.3 | XP_004008444.1 | 2 - 3 | 317 | 76 | |
| XM_012177642.3 | XP_012033032.1 | 2 - 3 | 259 | 76 | ||
| XM_027969323.1 | XP_027825124.1 | 2 - 3 | 626 | 76 | ||
| XM_027969324.1 | XP_027825125.1 | 2 - 3 | 262 | 76 | ||
| XM_027969325.1 | XP_027825126.1 | 2 - 3 | 268 | 76 | ||
| XM_027969326.1 | XP_027825127.1 | 2 - 3 | 623 | 76 | ||
More Information
Set Membership: |
|

MRPL55
Genomic Map