Assay Details
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012580.4 | XP_004012629.2 | 4 - 5 | 592 | 58 | |
XM_027975103.1 | XP_027830904.1 | 3 - 4 | 453 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
