Assay Details
| Gene Symbol: ZBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014768.4 | XP_004014817.1 | 5 - 6 | 2921 | 79 | |
| XM_012188922.3 | XP_012044312.1 | 4 - 5 | 3044 | 79 | ||
| XM_012188923.3 | XP_012044313.1 | 4 - 5 | 3041 | 79 | ||
| XM_027977016.1 | XP_027832817.1 | 5 - 6 | 1920 | 79 | ||
More Information
Set Membership: |
|

ZBP1
Genomic Map