Assay Details
| Gene Symbol: HAGHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961344.1 | XP_027817145.1 | 4 - 5 | 524 | 54 | |
| XM_027961345.1 | XP_027817146.1 | 4 - 5 | 615 | 54 | ||
| XM_027961346.1 | XP_027817147.1 | 4 - 5 | 489 | 54 | ||
| XM_027961347.1 | XP_027817148.1 | 4 - 5 | 412 | 54 | ||
| XM_027961348.1 | XP_027817149.1 | 4 - 5 | 528 | 54 | ||
| XM_027961349.1 | XP_027817150.1 | 4 - 5 | 522 | 54 | ||
| XM_027961350.1 | XP_027817151.1 | 4 - 5 | 608 | 54 | ||
| XM_027961352.1 | XP_027817153.1 | 4 - 5 | 424 | 54 | ||
| XM_027961353.1 | XP_027817154.1 | 4 - 5 | 523 | 54 | ||
| XM_027961354.1 | XP_027817155.1 | 4 - 5 | 436 | 54 | ||
| XM_027961355.1 | XP_027817156.1 | 4 - 5 | 569 | 54 | ||
More Information
Set Membership: |
|

HAGHL
Genomic Map