Assay Details
| Gene Symbol: NDUFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318974.2 | NP_001305903.2 | 1 | 237 | 82 | |
| XM_027971434.1 | XP_027827235.1 | 1 | 255 | 82 | ||
| GenBank mRNA | CD288450.1 | - | 264 | 82 | ||
| EE776519.1 | - | 1 | 245 | 82 | ||
More Information
Set Membership: |
|

NDUFA1
Genomic Map