Assay Details
Gene Symbol: LOC114112034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963710.1 | XP_027819511.1 | 2 - 3 | 363 | 70 |
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966512.1 | XP_027822313.1 | 7 - 8 | 1096 | 70 | |
XM_027966513.1 | XP_027822314.1 | 7 - 8 | 1096 | 70 |
Gene Symbol: LOC114112269 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963770.1 | XP_027819571.1 | 1 - 2 | 241 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
