Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019147.4 | XP_004019196.1 | 4 - 5 | 1082 | 73 | |
XM_027958633.1 | XP_027814434.1 | 4 - 5 | 466 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
