Assay Details
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957834.1 | XP_027813635.1 | 6 - 7 | 562 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957834.1 | XP_027813635.1 | 6 - 7 | 562 | 55 |
Set Membership: |
![]() ![]() ![]() |