Assay Details
Gene Symbol: TAS2R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012177256.2 | XP_012032646.1 | 1 | 1970 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
