Assay Details
Gene Symbol: CACNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973970.1 | XP_027829771.1 | 3 - 4 | 447 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
