Assay Details
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009990.4 | XP_004010039.1 | 3 - 4 | 352 | 80 | |
XM_027971181.1 | XP_027826982.1 | 3 - 4 | 483 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
