Assay Details
Gene Symbol: LOC114116372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973017.1 | XP_027828818.1 | 7 - 8 | 1033 | 96 | |
XM_027973018.1 | XP_027828819.1 | 7 - 8 | 1036 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
