Assay Details
| Gene Symbol: LOC101102085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977503.1 | XP_027833304.1 | 2 - 3 | 526 | 71 | |
| XM_027977504.1 | XP_027833305.1 | 2 - 3 | 528 | 71 | ||
| XM_027977506.1 | XP_027833307.1 | 2 - 3 | 527 | 71 | ||
More Information
Set Membership: |
|

LOC101102085
Genomic Map