Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 16 - 17 | 2419 | 80 | |
XM_012100634.2 | XP_011956024.1 | 12 - 13 | 1654 | 80 | ||
XM_027958477.1 | XP_027814278.1 | 15 - 16 | 2391 | 80 | ||
XM_027958478.1 | XP_027814279.1 | 17 - 18 | 2323 | 80 |
