Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 17 - 18 | 2548 | 71 | |
XM_012100634.2 | XP_011956024.1 | 13 - 14 | 1783 | 71 | ||
XM_027958477.1 | XP_027814278.1 | 16 - 17 | 2520 | 71 | ||
XM_027958478.1 | XP_027814279.1 | 18 - 19 | 2452 | 71 |
