Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 18 - 19 | 2807 | 88 | |
XM_012100634.2 | XP_011956024.1 | 14 - 15 | 2042 | 88 | ||
XM_027958477.1 | XP_027814278.1 | 17 - 18 | 2779 | 88 | ||
XM_027958478.1 | XP_027814279.1 | 19 - 20 | 2711 | 88 |
