Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 20 - 21 | 3203 | 93 | |
XM_012100634.2 | XP_011956024.1 | 16 - 17 | 2438 | 93 | ||
XM_027958477.1 | XP_027814278.1 | 19 - 20 | 3175 | 93 | ||
XM_027958478.1 | XP_027814279.1 | 21 - 22 | 3107 | 93 |
