Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 26 - 27 | 4191 | 87 | |
XM_012100634.2 | XP_011956024.1 | 22 - 23 | 3426 | 87 | ||
XM_027958477.1 | XP_027814278.1 | 25 - 26 | 4163 | 87 | ||
XM_027958478.1 | XP_027814279.1 | 27 - 28 | 4095 | 87 |
