Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 27 - 28 | 4302 | 102 | |
XM_012100634.2 | XP_011956024.1 | 23 - 24 | 3537 | 102 | ||
XM_027958477.1 | XP_027814278.1 | 26 - 27 | 4274 | 102 | ||
XM_027958478.1 | XP_027814279.1 | 28 - 29 | 4206 | 102 |

More Information
Set Membership: |
![]() ![]() |
