Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 29 - 30 | 4616 | 72 | |
XM_012100634.2 | XP_011956024.1 | 25 - 26 | 3851 | 72 | ||
XM_027958477.1 | XP_027814278.1 | 28 - 29 | 4588 | 72 | ||
XM_027958478.1 | XP_027814279.1 | 30 - 31 | 4520 | 72 |
