Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 6 - 7 | 939 | 94 | |
XM_012100634.2 | XP_011956024.1 | 2 - 3 | 174 | 94 | ||
XM_027958477.1 | XP_027814278.1 | 5 - 6 | 911 | 94 | ||
XM_027958478.1 | XP_027814279.1 | 7 - 8 | 843 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012100634.2 transcripts.
Set Membership: |
![]() ![]() |
