Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 7 - 8 | 1133 | 98 | |
XM_012100634.2 | XP_011956024.1 | 3 - 4 | 368 | 98 | ||
XM_027958477.1 | XP_027814278.1 | 6 - 7 | 1105 | 98 | ||
XM_027958478.1 | XP_027814279.1 | 8 - 9 | 1037 | 98 |
