Assay Details
Gene Symbol: TEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098945.2 | XP_014954431.1 | 3 - 4 | 380 | 76 | |
XM_015098946.2 | XP_014954432.1 | 3 - 4 | 381 | 76 | ||
XM_015098947.2 | XP_014954433.1 | 3 - 4 | 394 | 76 | ||
XM_027974105.1 | XP_027829906.1 | 3 - 4 | 607 | 76 |
