Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103726.2 | XP_014959212.2 | 3 - 4 | 1804 | 116 | |
| XM_027960896.1 | XP_027816697.1 | 2 - 3 | 4011 | 116 | ||
| XM_027960897.1 | XP_027816698.1 | 4 - 5 | 1543 | 116 | ||
More Information
Set Membership: |
|

ZBTB7C
Genomic Map