Assay Details
Gene Symbol: RPL36A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022487.3 | XP_004022536.1 | 1 - 2 | 137 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
