Assay Details
| Gene Symbol: LOC114108673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975165.1 | XP_027830966.1 | 5 - 6 | 617 | 90 | |
| Gene Symbol: LOC101113605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975163.1 | XP_027830964.1 | 4 - 5 | 527 | 90 | |

LOC114108673
Genomic Map