Assay Details
Gene Symbol: ELOVL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100862.3 | XP_011956252.2 | 7 - 8 | 908 | 82 | |
XM_027959009.1 | XP_027814810.1 | 7 - 8 | 1772 | 82 | ||
XM_027959010.1 | XP_027814811.1 | 5 - 6 | 1587 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
