Assay Details
| Gene Symbol: RAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974777.1 | XP_027830578.1 | 4 - 5 | 5917 | 82 | |
| XM_027974778.1 | XP_027830579.1 | 4 - 5 | 5791 | 82 | ||
| XM_027974779.1 | XP_027830580.1 | 2 - 3 | 5701 | 82 | ||
| XM_027974780.1 | XP_027830581.1 | 4 - 5 | 6734 | 82 | ||
More Information
Set Membership: |
|

RAI1
Genomic Map