Assay Details
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014778.3 | XP_004014827.2 | 1 - 2 | 79 | 92 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RSPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014778.3 | XP_004014827.2 | 1 - 2 | 79 | 92 |