Assay Details
| Gene Symbol: LOC101112990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973877.1 | XP_027829678.1 | 8 - 9 | 1052 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101112990| Gene Symbol: LOC101112990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973877.1 | XP_027829678.1 | 8 - 9 | 1052 | 100 | |
Set Membership: |
|