Assay Details
| Gene Symbol: NOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103961.3 | XP_011959351.2 | 4 - 5 | 724 | 64 | |
| XM_015104018.2 | XP_014959504.2 | 5 - 6 | 936 | 64 | ||
| XM_027961790.1 | XP_027817591.1 | 6 - 7 | 659 | 64 | ||
| XM_027961792.1 | XP_027817593.1 | 4 - 5 | 682 | 64 | ||
| XM_027961793.1 | XP_027817594.1 | 3 - 4 | 979 | 64 | ||
More Information
Set Membership: |
|

NOXO1
Genomic Map