Assay Details
| Gene Symbol: DISP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096942.2 | XP_014952428.2 | 4 - 5 | 597 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DISP2| Gene Symbol: DISP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096942.2 | XP_014952428.2 | 4 - 5 | 597 | 85 | |
Set Membership: |
|