Assay Details
| Gene Symbol: BID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968065.1 | XP_027823866.1 | 4 - 5 | 505 | 87 | |
| XM_027968066.1 | XP_027823867.1 | 4 - 5 | 585 | 87 | ||
More Information
Set Membership: |
|

BID
Genomic Map