Assay Details
| Gene Symbol: GTF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002169.3 | XP_004002218.1 | 4 - 5 | 517 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GTF2B| Gene Symbol: GTF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002169.3 | XP_004002218.1 | 4 - 5 | 517 | 75 | |
Set Membership: |
|