Assay Details
Gene Symbol: RHNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094992.2 | XP_014950478.1 | 2 - 3 | 321 | 70 | |
XM_015094993.2 | XP_014950479.1 | 2 - 3 | 315 | 70 | ||
XM_027968004.1 | XP_027823805.1 | 2 - 3 | 278 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
