Assay Details
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008729.4 | XP_004008778.2 | 4 - 5 | 466 | 112 | |
XM_027970046.1 | XP_027825847.1 | 4 - 5 | 1303 | 112 | ||
XM_027970047.1 | XP_027825848.1 | 4 - 5 | 509 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
