Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011743.3 | XP_004011792.1 | 4 | 1495 | 100 | |
| XM_004011744.3 | XP_004011793.1 | 4 | 1656 | 100 | ||
More Information
Set Membership: |
|

ELOC
Genomic Map