Assay Details
Gene Symbol: TARDBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975644.1 | XP_027831445.1 | 2 | 285 | 89 | |
XM_027975645.1 | XP_027831446.1 | 2 | 285 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
